missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2,2-Dimethylcyclopropanecarboxylic Acid 98.0+%, TCI America™

Catalog No. D36771G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36771G 1 g
D36775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36771G Supplier TCI America Supplier No. D36771G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14590-53-5
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD08460918
InChI Key BFNMOMYTTGHNGJ-SCSAIBSYSA-N
PubChem CID 10192561
IUPAC Name (1S)-2,2-dimethylcyclopropane-1-carboxylic acid
SMILES CC1(CC1C(=O)O)C

Specifications

Color Yellow
Boiling Point 100°C
Quantity 1 g
Formula Weight 114.14
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (S)-(+)-2,2-Dimethylcyclopropanecarboxylic Acid
EINECSNumber (3)-3774
TSCA No