missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2,2-Dimethylcyclopropanecarboxamide 98.0+%, TCI America™

Catalog No. p-7127856
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36765G 5 g
D367625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36765G Supplier TCI America Supplier No. D36765G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75885-58-4
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00216614
InChI Key YBZQRYWKYBZZNT-UHFFFAOYNA-N
Synonym s-+-2,2-dimethylcyclopropanecarboxamide, s-2,2-dimethylcyclopropanecarboxamide, s-+-2,2-dimethylcyclopropane carboxamide, s-+-2,2,-dimethylcyclopropane carboxamide, s-r-+-2,2-dimethyl cyclopropane carboxamide, s-+-2,2-dimethylcyclo-propane carboxamide, 1s-2,2-dimethylcyclopropanecarboxamide, 1s-2,2-dimethylcyclopropane-1-carboxamide, cyclopropanecarboxamide, 2,2-dimethyl-, 1s, s-2,2-dimethyl-cyclopropanecarboxylic acid amide
PubChem CID 10197657
IUPAC Name 2,2-dimethylcyclopropane-1-carboxamide
SMILES CC1(C)CC1C(N)=O

Specifications

Melting Point 136°C
Color White
Quantity 5 g
Formula Weight 113.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(+)-2,2-Dimethylcyclopropanecarboxamide
EINECSNumber (3)-3775
TSCA No