missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2,2-Dimethyl-1,3-dioxolane-4-methanol, 98%

Catalog No. p-3736309
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC218070050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC218070050 Supplier Thermo Scientific Chemicals Supplier No. 218070050
Only null left

CAS: 22323-82-6 | C6H12O3 | 132.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22323-82-6
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00063239
InChI Key RNVYQYLELCKWAN-UHFFFAOYNA-N
Synonym s-+-2,2-dimethyl-1,3-dioxolane-4-methanol, s-glycerol acetonide, s-+-1,2-isopropylideneglycerol, 4s-2,2-dimethyl-1,3-dioxolan-4-yl methanol, s-2,2-dimethyl-1,3-dioxolane-4-methanol, s-solketal, 1,2-isopropylidene-sn-glycerol, s-+-2,3-o-isopropylideneglycerol, 1,3-dioxolane-4-methanol, 2,2-dimethyl-, 4s, s-2,2-dimethyl-1,3-dioxolan-4-yl methanol
PubChem CID 736057
IUPAC Name [(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol
SMILES CC1(C)OCC(CO)O1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0700g/mL
Boiling Point 82.0°C to 83.0°C (14.0 mmHg)
Flash Point 80°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4330 to 1.4350
Quantity 5 g
Beilstein 19,III,647
Merck Index 10,3236
Specific Gravity 1.07
Specific Rotation Condition + 11.00 (20.00°C c=5,CH3OH)
Specific Rotation + 11.00
Solubility Information Solubility in water: miscible
Formula Weight 132.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material (S)-(+)-2, 2-Dimethyl-1, 3-dioxolane-4-methanol
Show More Show Less
EINECSNumber 244-910-8

RUO – Research Use Only