missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-2,2'-Dimethyl-1,1'-binaphthyl 98.0+%, TCI America™
$479.36 - $1619.07
Chemical Identifiers
| CAS | 32587-64-7 |
|---|---|
| Molecular Formula | C14H14 |
| Molecular Weight (g/mol) | 182.266 |
| MDL Number | MFCD00048075 |
| InChI Key | ABMKWMASVFVTMD-UHFFFAOYSA-N |
| PubChem CID | 11797 |
| IUPAC Name | 1-methyl-2-(2-methylphenyl)benzene |
| SMILES | CC1=CC=CC=C1C2=CC=CC=C2C |
Chemical Identifiers
| 32587-64-7 | |
| 182.266 | |
| ABMKWMASVFVTMD-UHFFFAOYSA-N | |
| 1-methyl-2-(2-methylphenyl)benzene |
| C14H14 | |
| MFCD00048075 | |
| 11797 | |
| CC1=CC=CC=C1C2=CC=CC=C2C |
Specifications
| 32587-64-7 | |
| MFCD00048075 | |
| ABMKWMASVFVTMD-UHFFFAOYSA-N | |
| 1-methyl-2-(2-methylphenyl)benzene | |
| 11797 | |
| ≥98.0% (HPLC) | |
| (S)-2,2′-Dimethyl-1,1′-binaphthyl |
| C14H14 | |
| 1 g | |
| CC1=CC=CC=C1C2=CC=CC=C2C | |
| 182.266 | |
| 282.39 | |
| Crystalline Powder |
Safety and Handling
TSCA : No