missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-(1-Hydroxyethyl)pyridine

Catalog No. AAJ6098003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6098003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6098003 Supplier Thermo Scientific Chemicals Supplier No. J6098003

CAS: 59042-90-9 | C7H9NO | 123.16 g/mol

Substituted 2-Pyridinemethanol derivatives are the potent and selective phosphodiesterase-4 inhibitors. Reaction of hydrous nitrosylruthenium trichloride with 2-pyridinemethanol gave ruthenium complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Substituted 2-Pyridinemethanol derivatives are the potent and selective phosphodiesterase-4 inhibitors. Reaction of hydrous nitrosylruthenium trichloride with 2-pyridinemethanol gave ruthenium complexes.

Solubility

Chemical Identifiers

CAS 59042-90-9
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.16
MDL Number MFCD06795465
InChI Key PPHIIIRFJKDTLG-UHFFFAOYNA-N
Synonym s-1-pyridin-2-yl ethanol, s-1-2-pyridyl ethanol, 2-s-1-hydroxyethyl pyridine, s---2-1-hydroxyethyl pyridine, s-2-1-hydroxyethyl pyridine, 1s-1-pyridin-2-yl ethan-1-ol, 1s-1-pyridin-2-yl ethanol, s-1-2-pyridyl ethanol, s-2-1-hydroxyethyl pyridine, pubchem5701, 1s-1-pyridin-2-ylethanol
PubChem CID 11094597
IUPAC Name (1S)-1-pyridin-2-ylethanol
SMILES CC(O)C1=CC=CC=N1

Specifications

Color Colorless
Quantity 1 g
Solubility Information Soluble in toluene.
Formula Weight 123.16
Physical Form Solid
Chemical Name or Material (S)-2-(1-Hydroxyethyl)pyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only