missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-(tert-Butoxycarbonyl)-4,4-difluoro-2-pyrrolidinecarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7123406
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4202200MG 200 mg
B42021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4202200MG Supplier TCI America Supplier No. B4202200MG

Chemical Identifiers

CAS 203866-15-3
Molecular Formula C10H15F2NO4
Molecular Weight (g/mol) 251.23
MDL Number MFCD03094917
InChI Key WTMZYKCXBXPVPT-UHFFFAOYNA-N
Synonym n-boc-4,4-difluoro-l-proline, s-1-tert-butoxycarbonyl-4,4-difluoropyrrolidine-2-carboxylic acid, n-t-boc-4,4-difluoro-l-proline, 1-tert-butoxycarbonyl-4,4-difluoro-l-proline, 2s-1-tert-butoxycarbonyl-4,4-difluoropyrrolidine-2-carboxylic acid, 2s-1-tert-butoxy carbonyl-4,4-difluoropyrrolidine-2-carboxylic acid, boc-4,4-difluoro-l-proline, s-1-boc-4,4-difluoropyrrolidine-2-carboxylic acid, n-boc-4,4-difluoroproline
PubChem CID 22977458
IUPAC Name 1-[(tert-butoxy)carbonyl]-4,4-difluoropyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CC(F)(F)CC1C(O)=O

Specifications

Melting Point 122°C
Quantity 200 mg
Formula Weight 251.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-1-(tert-Butoxycarbonyl)-4,4-difluoro-2-pyrrolidinecarboxylic Acid