missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-(tert-Butoxycarbonyl)-3-hydroxypiperidine 98.0+%, TCI America™

Catalog No. B44461G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44461G 1 g
B44465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44461G Supplier TCI America Supplier No. B44461G

Chemical Identifiers

CAS 143900-44-1
Molecular Formula C10H19NO3
Molecular Weight (g/mol) 201.27
MDL Number MFCD04115307
InChI Key UIJXHKXIOCDSEB-UHFFFAOYNA-N
Synonym s-1-boc-3-hydroxypiperidine, s-tert-butyl 3-hydroxypiperidine-1-carboxylate, s-n-boc-3-hydroxypiperidine, tert-butyl 3s-3-hydroxypiperidine-1-carboxylate, s-1-n-boc-3-hydroxy-piperidine, s-1-n-boc-3-hydroxypiperidine, s-n-boc-3-hydroxy piperidine, s-3-hydroxypiperidine, n-boc protected, s-3-hydroxy-piperidine-1-carboxylic acid tert-butyl ester, 3s-1-tert-butoxycarbonyl-3-hydroxypiperidine
PubChem CID 1514399
IUPAC Name tert-butyl 3-hydroxypiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(O)C1

Specifications

Melting Point 54°C
Color White
Quantity 1 g
Formula Weight 201.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-1-(tert-Butoxycarbonyl)-3-hydroxypiperidine