missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-(tert-Butoxycarbonyl)-2-piperidinecarboxylic Acid 98.0+%, TCI America™

Catalog No. B37001G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37001G 1 g
B37005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37001G Supplier TCI America Supplier No. B37001G

Chemical Identifiers

CAS 26250-84-0
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.276
MDL Number MFCD00151904
InChI Key JQAOHGMPAAWWQO-QMMMGPOBSA-N
Synonym boc-pip-oh, s-1-boc-piperidine-2-carboxylic acid, s-n-boc-piperidine-2-carboxylic acid, s-1-tert-butoxycarbonyl piperidine-2-carboxylic acid, boc-l-pipecolic acid, boc-l-pip-oh, n-boc-l-pipecolinic acid, l-n-boc-pipecolic acid, 2s-1-tert-butoxycarbonyl piperidine-2-carboxylic acid, 2s-1-tert-butoxy carbonyl piperidine-2-carboxylic acid
PubChem CID 688617
IUPAC Name (2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCCCC1C(=O)O

Specifications

Melting Point 124°C
Color White
Quantity 1 g
Formula Weight 229.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-1-(tert-Butoxycarbonyl)-2-piperidinecarboxylic Acid