missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-(tert-Butoxycarbonyl)-2-methylpiperazine 98.0+%, TCI America™

Catalog No. B48461G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48461G 1 g
B48465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48461G Supplier TCI America Supplier No. B48461G
Only null left

Chemical Identifiers

CAS 169447-70-5
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD01862121
InChI Key DATRVIMZZZVHMP-QMMMGPOBSA-N
Synonym (S)-1-Boc-2-methylpiperazine, tert-Butyl (S)-2-Methyl-1-piperazinecarboxylate, (S)-2-Methyl-1-piperazinecarboxylic Acid tert-Butyl Ester
PubChem CID 10081508
IUPAC Name tert-butyl (2S)-2-methylpiperazine-1-carboxylate
SMILES CC1CNCCN1C(=O)OC(C)(C)C

Specifications

Melting Point 45°C
Color White
Quantity 1 g
Formula Weight 200.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-1-(tert-Butoxycarbonyl)-2-methylpiperazine