missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-(tert-Butoxycarbonyl)-2-cyanopyrrolidine 96.0+%, TCI America™

Catalog No. B48781G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B48781G Supplier TCI America Supplier No. B48781G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 228244-04-0
Melting Point 33°C
Color White-Yellow
Molecular Formula C10H16N2O2
Quantity 1 g
MDL Number MFCD01861224
Synonym s-1-boc-2-cyanopyrrolidine, s-tert-butyl 2-cyanopyrrolidine-1-carboxylate, s-1-n-boc-2-cyano-pyrrolidine, tert-butyl 2s-2-cyanopyrrolidine-1-carboxylate, s-1-boc-2-cyano-pyrrolidine, s-1-n-boc-2-cyanopyrrolidine, s-1-boc-2-pyrrolidinecarbonitrile, s-n-boc-pyrrolidine-2-carbonitrile, s---1-boc-2-cyanopyrrolidine, s-n-boc-2-pyrrolidinonitrile
InChI Key MDMSZBHMBCNYNO-UHFFFAOYNA-N
SMILES CC(C)(C)OC(=O)N1CCCC1C#N
IUPAC Name tert-butyl 2-cyanopyrrolidine-1-carboxylate
Molecular Weight (g/mol) 196.25
PubChem CID 6951263
Formula Weight 196.25
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material (S)-1-(tert-Butoxycarbonyl)-2-cyanopyrrolidine
Show More Show Less
TSCA No
Recommended Storage Refrigerator