missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-Phenylethyl isocyanate, 98%

Catalog No. AAL1114103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1114103 1 g
AAL1114106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1114103 Supplier Thermo Scientific Chemicals Supplier No. L1114103

CAS: 14649-03-7 | C9H9NO | 147.177 g/mol

(S)-(-)-1-Phenylethyl isocyanate act by inhibition of cell proliferation and induction of apoptosis. It helps to prevent lung cancer and esophageal cancer. It is a fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-1-Phenylethyl isocyanate act by inhibition of cell proliferation and induction of apoptosis. It helps to prevent lung cancer and esophageal cancer. It is a fine chemical intermediate.

Solubility
Hydrolyzes in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive to moisture. Recommended storage temperature is 2 - 8°C.

Chemical Identifiers

CAS 14649-03-7
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00002036
InChI Key JJSCUXAFAJEQGB-QMMMGPOBSA-N
Synonym s---1-phenylethyl isocyanate, 1s-1-isocyanatoethyl benzene, s---alpha-methylbenzyl isocyanate, benzene, 1s-1-isocyanatoethyl, isocyanic acid s---a-methylbenzyl ester, s-alpha-methylbenzyl isocyanate, s---1-phenylethylisocyanate, isocyanic acid s---alpha-methylbenzyl ester, isocyanic acid s---methylbenzyl ester, 1-isocyanatoethyl benzene #
PubChem CID 2724173
IUPAC Name [(1S)-1-isocyanatoethyl]benzene
SMILES CC(C1=CC=CC=C1)N=C=O

Specifications

Density 1.045
Boiling Point 55°C to 56°C (2.5 mmHg)
Flash Point 65°C (149°F)
Odor Pungent
Refractive Index 1.5145
Quantity 1 g
UN Number UN2206
Beilstein 4230972
Sensitivity Moisture sensitive; May form a precipitate
Solubility Information Hydrolyzes in water.
Optical Rotation −10.5° (Neat)
Formula Weight 147.18
Percent Purity 98%
Chemical Name or Material (S)-(-)-1-Phenylethyl isocyanate
Show More Show Less
Hazard Category H227-H315-H319-H330-H334-H335
Hazard Statement GHS H Statement
H331-H334-H227-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P284-P285-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 238-698-6
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only