missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-Phenyl-2-(p-tolyl)ethylamine 98.0+%, TCI America™

Catalog No. P11185G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11185G 5 g
P111825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11185G Supplier TCI America Supplier No. P11185G

Chemical Identifiers

CAS 30339-30-1
Molecular Formula C15H17N
Molecular Weight (g/mol) 211.31
MDL Number MFCD00221505
InChI Key ZICDZTXDTPZBKH-UHFFFAOYNA-N
PubChem CID 10160531
IUPAC Name 2-(4-methylphenyl)-1-phenylethan-1-amine
SMILES CC1=CC=C(CC(N)C2=CC=CC=C2)C=C1

Specifications

Quantity 5 g
Formula Weight 211.31
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (S)-1-Phenyl-2-(p-tolyl)ethylamine