missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-Methyl-3-pyrrolidinol 98.0+%, TCI America™

Catalog No. p-7132923
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19381G 1 g
M19385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19381G Supplier TCI America Supplier No. M19381G

Chemical Identifiers

CAS 104641-59-0
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD03788748
InChI Key FLVFPAIGVBQGET-UHFFFAOYNA-N
Synonym s-+-1-methyl-3-pyrrolidinol, 3s-1-methylpyrrolidin-3-ol, s-1-methyl-3-pyrrolidinol, s-1-methylpyrrolidin-3-ol, 3-pyrrolidinol, 1-methyl-, 3s, s-+-3-hydroxy-n-methylpyrrolidine, s-+-1-methyl-3-hydroxypyrrolidine, s-3-hydroxy-1-methylpyrrolidine, 1-methyl-3-pyrrolidinol, +, s-3-hydroxy-1-methyl-pyrrolidine
PubChem CID 643504
IUPAC Name 1-methylpyrrolidin-3-ol
SMILES CN1CCC(O)C1

Specifications

Color Yellow
Boiling Point 182°C
Quantity 1 g
Formula Weight 101.15
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (S)-1-Methyl-3-pyrrolidinol