missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1-Methyl-3-pyrrolidinol, 98%, ee 99%

CAS: 104641-59-0 | C5H11NO | 101.15 g/mol

$205.12 - $205.12

Chemical Identifiers

CAS 104641-59-0
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD03788748
InChI Key FLVFPAIGVBQGET-UHFFFAOYNA-N
Synonym s-+-1-methyl-3-pyrrolidinol, 3s-1-methylpyrrolidin-3-ol, s-1-methyl-3-pyrrolidinol, s-1-methylpyrrolidin-3-ol, 3-pyrrolidinol, 1-methyl-, 3s, s-+-3-hydroxy-n-methylpyrrolidine, s-+-1-methyl-3-hydroxypyrrolidine, s-3-hydroxy-1-methylpyrrolidine, 1-methyl-3-pyrrolidinol, +, s-3-hydroxy-1-methyl-pyrrolidine
PubChem CID 643504
SMILES CN1CCC(O)C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC439320010
SDS View Documents
Thermo Scientific Chemicals
439320010
1 g
Each for $205.12
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

104641-59-0
101.15
FLVFPAIGVBQGET-UHFFFAOYNA-N
643504
C5H11NO
MFCD03788748
s-+-1-methyl-3-pyrrolidinol, 3s-1-methylpyrrolidin-3-ol, s-1-methyl-3-pyrrolidinol, s-1-methylpyrrolidin-3-ol, 3-pyrrolidinol, 1-methyl-, 3s, s-+-3-hydroxy-n-methylpyrrolidine, s-+-1-methyl-3-hydroxypyrrolidine, s-3-hydroxy-1-methylpyrrolidine, 1-methyl-3-pyrrolidinol, +, s-3-hydroxy-1-methyl-pyrrolidine
CN1CCC(O)C1

Specifications

104641-59-0
100.0
0.9930g/mL
90°C
97.5% min. (GC)
MFCD03788748
0.993
s-+-1-methyl-3-pyrrolidinol, 3s-1-methylpyrrolidin-3-ol, s-1-methyl-3-pyrrolidinol, s-1-methylpyrrolidin-3-ol, 3-pyrrolidinol, 1-methyl-, 3s, s-+-3-hydroxy-n-methylpyrrolidine, s-+-1-methyl-3-hydroxypyrrolidine, s-3-hydroxy-1-methylpyrrolidine, 1-methyl-3-pyrrolidinol, +, s-3-hydroxy-1-methyl-pyrrolidine
CN1CCC(O)C1
101.15
101.15
Liquid
97.5
Brown to Colorless
181°C to 182°C
Authentic
C5H11NO
1 g
7
FLVFPAIGVBQGET-UHFFFAOYNA-N
(3S)-1-methylpyrrolidin-3-ol
643504
98%,ee99%
(S)-(+)-1-Methyl-3-pyrrolidinol, ee 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Recommended Storage : May darken on storage with no loss of purity

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only