Learn More
(S)-(+)-1-Methoxy-2-propanol, 99%
CAS: 26550-55-0 | C4H10O2 | 90.12 g/mol
$754.56 - $754.56
Chemical Identifiers
| CAS | 26550-55-0 |
|---|---|
| Molecular Formula | C4H10O2 |
| Molecular Weight (g/mol) | 90.12 |
| MDL Number | MFCD01632588 |
| InChI Key | ARXJGSRGQADJSQ-BYPYZUCNSA-N |
| Synonym | s-+-1-methoxy-2-propanol, 2s-1-methoxypropan-2-ol, s-1-methoxypropan-2-ol, s---1-methoxy-2-propanol, s-1-methoxy-2-propanol, 2-propanol, 1-methoxy-, 2s, 2s-1-methoxy-2-propanol, pubchem6715, s-1-methoxy-propan-2-ol |
| PubChem CID | 6993459 |
| IUPAC Name | (2S)-1-methoxypropan-2-ol |
| SMILES | CC(COC)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC331790050
|
Thermo Scientific Chemicals
331790050 |
5 mL | Glass bottle |
Each for $754.56
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 26550-55-0 | |
| 90.12 | |
| ARXJGSRGQADJSQ-BYPYZUCNSA-N | |
| 6993459 | |
| CC(COC)O |
| C4H10O2 | |
| MFCD01632588 | |
| s-+-1-methoxy-2-propanol, 2s-1-methoxypropan-2-ol, s-1-methoxypropan-2-ol, s---1-methoxy-2-propanol, s-1-methoxy-2-propanol, 2-propanol, 1-methoxy-, 2s, 2s-1-methoxy-2-propanol, pubchem6715, s-1-methoxy-propan-2-ol | |
| (2S)-1-methoxypropan-2-ol |
Specifications
| 26550-55-0 | |
| 100.0 | |
| Colorless | |
| 120.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4020 to 1.4040 | |
| MFCD01632588 | |
| 0.92 | |
| + 20.00 | |
| Solubility in water: soluble | |
| CC(COC)O | |
| 90.12 | |
| 90.12 | |
| Liquid |
| 98.5 | |
| -97.0°C | |
| 0.9200g/mL | |
| 30°C | |
| 98.5% min. (GC) | |
| C4H10O2 | |
| CH3OCH2CH(OH)CH3 | |
| 5 mL | |
| +20° (20°C c=10,CHCl3) | |
| s-+-1-methoxy-2-propanol, 2s-1-methoxypropan-2-ol, s-1-methoxypropan-2-ol, s---1-methoxy-2-propanol, s-1-methoxy-2-propanol, 2-propanol, 1-methoxy-, 2s, 2s-1-methoxy-2-propanol, pubchem6715, s-1-methoxy-propan-2-ol | |
| ARXJGSRGQADJSQ-BYPYZUCNSA-N | |
| (2S)-1-methoxypropan-2-ol | |
| 6993459 | |
| 99% | |
| (S)-(+)-1-Methoxy-2-propanol |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
May cause drowsiness or dizziness.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Avoid breathing dust/fume/gas/mist/vapors/spray.
GHS Signal Word: Warning
RUO – Research Use Only