missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-Ethyl-2-pyrrolidinecarboxamide 98.0+%, TCI America™

Catalog No. E04521G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04521G 1 g
E04525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E04521G Supplier TCI America Supplier No. E04521G

Chemical Identifiers

CAS 114812-34-9
Molecular Formula C7H14N2O
Molecular Weight (g/mol) 142.202
MDL Number MFCD00191361
InChI Key LQOASEHNECNLNM-LURJTMIESA-N
Synonym N-Ethyl-L-prolinamide
PubChem CID 10866443
IUPAC Name (2S)-1-ethylpyrrolidine-2-carboxamide
SMILES CCN1CCCC1C(=O)N

Specifications

Melting Point 110°C
Color White-Yellow
Quantity 1 g
Formula Weight 142.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-1-Ethyl-2-pyrrolidinecarboxamide