missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-Cyclopropylethylamine, ChiPros™, 98%, ee 98+%

Catalog No. AAH2749906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2749906 5 g
AAH2749903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2749906 Supplier Thermo Scientific Chemicals Supplier No. H2749906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 195604-39-8 | C5H11N | 85.15 g/mol

(S)-1-Cyclopropylethylamine acts as a chiral and organic building block in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-1-Cyclopropylethylamine acts as a chiral and organic building block in organic synthesis.

Solubility
Miscible with water.

Notes
Air sensitive. Incompatible with acids and strong oxidizing agents.

Chemical Identifiers

CAS 195604-39-8
Molecular Formula C5H11N
Molecular Weight (g/mol) 85.15
MDL Number MFCD08064289
InChI Key IXCXVGWKYIDNOS-BYPYZUCNSA-N
PubChem CID 7363995
IUPAC Name (1S)-1-cyclopropylethanamine
SMILES CC(C1CC1)N

Specifications

Melting Point -68°C
Density 0.799
Boiling Point 95°C
Flash Point 3°C (37°F)
Quantity 5 g
UN Number UN2733
Sensitivity Air Sensitive
Synonym s-1-cyclopropylethylamine, s-1-cyclopropylethanamine, 1s-1-cyclopropylethanamine, 1s-1-cyclopropylethan-1-amine, 1 s-1-cyclopropylethanamine, n-s-1-cyclopropyl-ethyl-amine, s-1-cyclopropylethylamine, chipros, cyclopropanemethanamine,a-methyl-, as, s-1-cyclopropylethylamine, chipros, ee 98+%, s-1-cyclopropylethylamine, chipros r , produced by basf
Solubility Information Miscible with water.
Formula Weight 85.15
Percent Purity 98%
Assay ee ≥98%
Chemical Name or Material (S)-1-Cyclopropylethylamine
Show More Show Less
Hazard Category H225-H314-H335
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only