missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1-Chloro-2-methylbutane 98.0+%, TCI America™

$186.41 - $318.75

Chemical Identifiers

CAS 40560-29-0
Molecular Formula C5H11Cl
Molecular Weight (g/mol) 106.59
MDL Number MFCD00221494
InChI Key IWAKWOFEHSYKSI-UHFFFAOYNA-N
PubChem CID 12431745
IUPAC Name 1-chloro-2-methylbutane
SMILES CCC(C)CCl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C118810ML
View Documents
TCI America
C118810ML
10 mL
Each for $186.41
Only null left
 
C118825ML
View Documents
TCI America
C118825ML
25 mL
Each for $318.75
Only null left
 
Description

Description

Specifications

Chemical Identifiers

40560-29-0
106.59
IWAKWOFEHSYKSI-UHFFFAOYNA-N
1-chloro-2-methylbutane
C5H11Cl
MFCD00221494
12431745
CCC(C)CCl

Specifications

40560-29-0
100°C
MFCD00221494
1107
CCC(C)CCl
106.59
106.59
Liquid
Colorless
C5H11Cl
10 mL
IWAKWOFEHSYKSI-UHFFFAOYNA-N
1-chloro-2-methylbutane
12431745
≥98.0% (GC)
(S)-(+)-1-Chloro-2-methylbutane
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0066

TSCA : No

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'