missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1-Chloro-2-methylbutane 98.0+%, TCI America™

Catalog No. C118810ML
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C118810ML 10 mL
C118825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C118810ML Supplier TCI America Supplier No. C118810ML
Only null left

Chemical Identifiers

CAS 40560-29-0
Molecular Formula C5H11Cl
Molecular Weight (g/mol) 106.59
MDL Number MFCD00221494
InChI Key IWAKWOFEHSYKSI-UHFFFAOYNA-N
PubChem CID 12431745
IUPAC Name 1-chloro-2-methylbutane
SMILES CCC(C)CCl

Specifications

Color Colorless
Boiling Point 100°C
Quantity 10 mL
UN Number 1107
Formula Weight 106.59
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-(+)-1-Chloro-2-methylbutane
EINECSNumber (2)-0066
TSCA No
Recommended Storage Refrigerator