missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-(Boc-amino)-2-propanol, 97%

Catalog No. AAH5282606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5282606 5 g
AAH5282614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5282606 Supplier Thermo Scientific Chemicals Supplier No. H5282606

CAS: 167938-56-9 | C8H17NO3 | 175.228 g/mol

(S)-1-(Boc-amino)-2-propanol is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-1-(Boc-amino)-2-propanol is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

Chemical Identifiers

CAS 167938-56-9
Molecular Formula C8H17NO3
Molecular Weight (g/mol) 175.228
MDL Number MFCD04974340
InChI Key YNJCFDAODGKHAV-LURJTMIESA-N
Synonym n-boc-s-1-amino-2-propanol, s-tert-butyl 2-hydroxypropyl carbamate, s-1-boc-amino-2-propanol, tert-butyl n-2s-2-hydroxypropyl carbamate, tert-butyl 2s-2-hydroxypropyl carbamate, s-2-hydroxy-propyl-carbamic acid t-butyl ester, s-2-hydroxypropyl carbamic acid tert-butyl ester, s-2-hydroxy-propyl-carbamic acid tert-butyl ester
PubChem CID 11789722
IUPAC Name tert-butyl N-[(2S)-2-hydroxypropyl]carbamate
SMILES CC(CNC(=O)OC(C)(C)C)O

Specifications

Quantity 5 g
Formula Weight 175.23
Percent Purity 97%
Chemical Name or Material (S)-1-(Boc-amino)-2-propanol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only