missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-Boc-4-oxopiperidine-2-carboxylic acid, 95%, Thermo Scientific Chemicals

Catalog No. p-7038611
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5761703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5761703 Supplier Thermo Scientific Chemicals Supplier No. H5761703

It is a fine chemical and synthetic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a fine chemical and synthetic intermediate.

Solubility
Soluble (12 g/L) (25°C)

Notes
Store in a cool ventilated place. Avoid strong oxidizing materials. Hygroscopic.

Chemical Identifiers

CAS 198646-60-5
Molecular Formula C11H17NO5
Molecular Weight (g/mol) 243.259
MDL Number MFCD03094867
InChI Key GPBCBXYUAJQMQM-QMMMGPOBSA-N
Synonym s-1-tert-butoxycarbonyl-4-oxopiperidine-2-carboxylic acid, s-1-boc-4-oxopipecolic acid, s---1-boc-4-oxopiperidine-2-carboxylic acid, s-1-boc-4-oxopiperidine-2-carboxylic acid, 2s-1-tert-butoxycarbonyl-4-oxopiperidine-2-carboxylic acid, 2s-1-tert-butoxy carbonyl-4-oxopiperidine-2-carboxylic acid, s-1-tert-butoxycarbonyl-4-oxopiperidine-2-carboxylicacid, 2s-4-oxo-1,2-piperidinedicarboxylic acid 1-1,1-dimethylethyl ester, 2s-n-boc-4-oxopipecolic acid, r-+-1-boc-4-oxopiperidine-2-carboxylic acid
PubChem CID 10857681
IUPAC Name (2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]-4-oxopiperidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCC(=O)CC1C(=O)O

Specifications

Melting Point 121°C to 126°C
Quantity 1 g
Sensitivity Air and moisture sensitive
Solubility Information Soluble (12g/L) (25°C)
Optical Rotation −22° (c=1 in Chloroform)
Formula Weight 243.26
Percent Purity 95%
Chemical Name or Material (S)-(-)-1-Boc-4-oxopiperidine-2-carboxylic acid
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only