Learn More
(S)-1-Boc-3-hydroxypiperidine, 97%
CAS: 143900-44-1 | C10H19NO3 | 201.27 g/mol
$104.32 - $428.97
Chemical Identifiers
| CAS | 143900-44-1 |
|---|---|
| Molecular Formula | C10H19NO3 |
| Molecular Weight (g/mol) | 201.27 |
| MDL Number | MFCD04115307 |
| InChI Key | UIJXHKXIOCDSEB-UHFFFAOYNA-N |
| Synonym | s-1-boc-3-hydroxypiperidine, s-tert-butyl 3-hydroxypiperidine-1-carboxylate, s-n-boc-3-hydroxypiperidine, tert-butyl 3s-3-hydroxypiperidine-1-carboxylate, s-1-n-boc-3-hydroxy-piperidine, s-1-n-boc-3-hydroxypiperidine, s-n-boc-3-hydroxy piperidine, s-3-hydroxypiperidine, n-boc protected, s-3-hydroxy-piperidine-1-carboxylic acid tert-butyl ester, 3s-1-tert-butoxycarbonyl-3-hydroxypiperidine |
| PubChem CID | 1514399 |
| IUPAC Name | tert-butyl 3-hydroxypiperidine-1-carboxylate |
| SMILES | CC(C)(C)OC(=O)N1CCCC(O)C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5277103
|
Thermo Scientific Chemicals
H5277103 |
1 g |
Each for $104.32
|
|
|||||
|
AAH5277106
|
Thermo Scientific Chemicals
H5277106 |
5 g |
Each for $428.97
|
|
|||||
Description
(S)-1-Boc-3-hydroxypiperidine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(S)-1-Boc-3-hydroxypiperidine is used as pharmaceutical intermediate.
Solubility
Soluble in methanol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
| 143900-44-1 | |
| 201.27 | |
| UIJXHKXIOCDSEB-UHFFFAOYNA-N | |
| 1514399 | |
| CC(C)(C)OC(=O)N1CCCC(O)C1 |
| C10H19NO3 | |
| MFCD04115307 | |
| s-1-boc-3-hydroxypiperidine, s-tert-butyl 3-hydroxypiperidine-1-carboxylate, s-n-boc-3-hydroxypiperidine, tert-butyl 3s-3-hydroxypiperidine-1-carboxylate, s-1-n-boc-3-hydroxy-piperidine, s-1-n-boc-3-hydroxypiperidine, s-n-boc-3-hydroxy piperidine, s-3-hydroxypiperidine, n-boc protected, s-3-hydroxy-piperidine-1-carboxylic acid tert-butyl ester, 3s-1-tert-butoxycarbonyl-3-hydroxypiperidine | |
| tert-butyl 3-hydroxypiperidine-1-carboxylate |
Specifications
| 143900-44-1 | |
| >110°C (230°F) | |
| MFCD04115307 | |
| s-1-boc-3-hydroxypiperidine, s-tert-butyl 3-hydroxypiperidine-1-carboxylate, s-n-boc-3-hydroxypiperidine, tert-butyl 3s-3-hydroxypiperidine-1-carboxylate, s-1-n-boc-3-hydroxy-piperidine, s-1-n-boc-3-hydroxypiperidine, s-n-boc-3-hydroxy piperidine, s-3-hydroxypiperidine, n-boc protected, s-3-hydroxy-piperidine-1-carboxylic acid tert-butyl ester, 3s-1-tert-butoxycarbonyl-3-hydroxypiperidine | |
| UIJXHKXIOCDSEB-UHFFFAOYNA-N | |
| CC(C)(C)OC(=O)N1CCCC(O)C1 | |
| 201.27 | |
| 201.27 | |
| (S)-1-Boc-3-hydroxypiperidine |
| 46°C to 50°C | |
| C10H19NO3 | |
| 1 g | |
| Soluble in methanol. | |
| +23° (c=5 in Methanol) | |
| tert-butyl 3-hydroxypiperidine-1-carboxylate | |
| 1514399 | |
| 97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only