missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1-Boc-3-bromopyrrolidine, 95%

Catalog No. p-7038447
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH57408MD 250 mg
AAH5740803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH57408MD Supplier Thermo Scientific Chemicals Supplier No. H57408MD

CAS: 569660-89-5 | C9H16BrNO2 | 250.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.

Chemical Identifiers

CAS 569660-89-5
Molecular Formula C9H16BrNO2
Molecular Weight (g/mol) 250.14
MDL Number MFCD17214728
InChI Key QJTKPXFJOXKUEY-UHFFFAOYNA-N
Synonym s-tert-butyl 3-bromopyrrolidine-1-carboxylate, s-3-bromo-pyrrolidine-1-carboxylic acid tert-butyl ester, tert-butyl 3s-3-bromopyrrolidine-1-carboxylate, s-+-1-boc-3-bromopyrrolidine, tert-butyl s-3-bromopyrrolidine-1-carboxylate, s-3-bromopyrrolidine-1-carboxylic acid tert-butyl ester
PubChem CID 34179480
IUPAC Name tert-butyl (3S)-3-bromopyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(Br)C1

Specifications

Quantity 250 mg
UN Number UN3077
Optical Rotation +33° (c=0.8 in Chloroform)
Formula Weight 250.14
Percent Purity 95%
Chemical Name or Material (S)-(+)-1-Boc-3-bromopyrrolidine
Hazard Category H302-H319
Hazard Statement GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only