missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-BOC-3-(Aminomethyl)piperidine, 97%, Thermo Scientific™

Catalog No. p-4228396
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC437470010 1g
AC437470050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC437470010 Supplier Thermo Scientific Chemicals Supplier No. 437470010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 140645-24-5
Molecular Formula C11H22N2O2
Molecular Weight (g/mol) 214.31
InChI Key WPWXYQIMXTUMJB-VIFPVBQESA-N
Synonym s-1-boc-3-aminomethyl piperidine, s-n-boc-3-aminomethylpiperidine, s-3-aminomethyl-1-boc-piperidine, s-tert-butyl 3-aminomethyl piperidine-1-carboxylate, s-1-n-boc-3-aminomethyl piperidine, s-3-aminomethyl-1-n-boc-piperidine, tert-butyl 3s-3-aminomethyl piperidine-1-carboxylate, s-3-aminomethyl-piperidine-1-carboxylic acid tert-butyl ester, s-1-n-boc-piperidine-3-methylamine, 3s-3-aminomethyl-n-tert-butoxycarbonylpiperidine
PubChem CID 1502022
IUPAC Name tert-butyl (3S)-3-(aminomethyl)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(C1)CN

Specifications

CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1g
Specific Rotation -15
Formula Weight 214.31
Percent Purity 97%
Chemical Name or Material (S)-1-BOC-3-(Aminomethyl)piperidine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only