missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-Boc-2-isopropylpiperazine, 97%

CAS: 674792-05-3 | C12H24N2O2 | 228.34 g/mol

Supplier:  Thermo Scientific Chemicals H5242403

 View more versions of this product

Catalog No. AAH5242403


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(S)-1-Boc-2-isopropylpiperazine
C12H24N2O2
1 g
s-1-boc-2-isopropylpiperazine, s-1-boc-2-isopropyl-piperazine, s-tert-butyl 2-isopropylpiperazine-1-carboxylate, tert-butyl 2s-2-propan-2-yl piperazine-1-carboxylate, 1-n-boc-2s-isopropyl-piperazine, tert-butyl 2s-2-isopropylpiperazine-1-carboxylate, s-n-1-boc-2-isopropylpiperazine, 2s-1-n-boc-2-isopropylpiperazine, s-1-n-boc-2-isopropyl piperazine
CC(C)[C@H]1CNCCN1C(=O)OC(C)(C)C
228.34
228.33
674792-05-3
MFCD03787918
Moisture sensitive
NZTWGWFHWJARJX-SNVBAGLBSA-N
tert-butyl (2S)-2-propan-2-ylpiperazine-1-carboxylate
17750439
97%

Chemical Identifiers

674792-05-3
228.34
NZTWGWFHWJARJX-SNVBAGLBSA-N
17750439
CC(C)[C@H]1CNCCN1C(=O)OC(C)(C)C
C12H24N2O2
MFCD03787918
s-1-boc-2-isopropylpiperazine, s-1-boc-2-isopropyl-piperazine, s-tert-butyl 2-isopropylpiperazine-1-carboxylate, tert-butyl 2s-2-propan-2-yl piperazine-1-carboxylate, 1-n-boc-2s-isopropyl-piperazine, tert-butyl 2s-2-isopropylpiperazine-1-carboxylate, s-n-1-boc-2-isopropylpiperazine, 2s-1-n-boc-2-isopropylpiperazine, s-1-n-boc-2-isopropyl piperazine
tert-butyl (2S)-2-propan-2-ylpiperazine-1-carboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only