missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-Boc-2-isopropylpiperazine, 97%

Catalog No. AAH52424MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52424MD 250 mg
AAH5242403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52424MD Supplier Thermo Scientific Chemicals Supplier No. H52424MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 674792-05-3 | C12H24N2O2 | 228.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 674792-05-3
Molecular Formula C12H24N2O2
Molecular Weight (g/mol) 228.34
MDL Number MFCD03787918
InChI Key NZTWGWFHWJARJX-SNVBAGLBSA-N
Synonym s-1-boc-2-isopropylpiperazine, s-1-boc-2-isopropyl-piperazine, s-tert-butyl 2-isopropylpiperazine-1-carboxylate, tert-butyl 2s-2-propan-2-yl piperazine-1-carboxylate, 1-n-boc-2s-isopropyl-piperazine, tert-butyl 2s-2-isopropylpiperazine-1-carboxylate, s-n-1-boc-2-isopropylpiperazine, 2s-1-n-boc-2-isopropylpiperazine, s-1-n-boc-2-isopropyl piperazine
PubChem CID 17750439
IUPAC Name tert-butyl (2S)-2-propan-2-ylpiperazine-1-carboxylate
SMILES CC(C)[C@H]1CNCCN1C(=O)OC(C)(C)C

Specifications

Quantity 250 mg
Sensitivity Moisture sensitive
Formula Weight 228.33
Percent Purity 97%
Chemical Name or Material (S)-1-Boc-2-isopropylpiperazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only