missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-Boc-2-isobutylpiperazine, 97%

Catalog No. AAH52409MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52409MD 250 mg
AAH5240903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52409MD Supplier Thermo Scientific Chemicals Supplier No. H52409MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 674792-06-4 | C13H26N2O2 | 242.363 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 674792-06-4
Molecular Formula C13H26N2O2
Molecular Weight (g/mol) 242.363
MDL Number MFCD03787916
InChI Key WXGGVOBNOVOVAM-NSHDSACASA-N
Synonym s-1-boc-2-isobutyl-piperazine, s-tert-butyl 2-isobutylpiperazine-1-carboxylate, s-1-boc-2-isobutylpiperazine, tert-butyl 2s-2-2-methylpropyl piperazine-1-carboxylate, s-1-n-boc-isobutylpiperazine, s-n-boc-2-isobutylpiperazine, tert-butyl 2s-2-isobutylpiperazine-1-carboxylate, s-1-n-boc-2-isobutylpiperazine, 2s-1-n-boc-2-isobutylpiperazine, 2s-1-tert-butoxycarbonyl-2-isobutylpiperazine
PubChem CID 17750437
IUPAC Name tert-butyl (2S)-2-(2-methylpropyl)piperazine-1-carboxylate
SMILES CC(C)CC1CNCCN1C(=O)OC(C)(C)C

Specifications

Quantity 250 mg
Formula Weight 242.36
Percent Purity 97%
Chemical Name or Material (S)-1-Boc-2-isobutylpiperazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only