missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-Amino-2-(methoxymethyl)pyrrolidine 98.0+%, TCI America™

Catalog No. p-7117375
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13351G 1 g
A13355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A13351G Supplier TCI America Supplier No. A13351G

Chemical Identifiers

CAS 59983-39-0
Molecular Formula C6H14N2O
Molecular Weight (g/mol) 130.191
MDL Number MFCD00064485
InChI Key BWSIKGOGLDNQBZ-LURJTMIESA-N
Synonym s---1-amino-2-methoxymethyl pyrrolidine, samp, s-2-methoxymethyl pyrrolidin-1-amine, 2s-2-methoxymethyl pyrrolidin-1-amine, 1-pyrrolidinamine, 2-methoxymethyl-, 2s, 2s-2-methoxymethyl-1-pyrrolidinamine, s-1-amino-2-methoxymethyl pyrrolidine, s---2-methoxymethyl-1-pyrrolidinamine, 2-methoxymethyl-1-pyrrolidinamine #
PubChem CID 7156993
IUPAC Name (2S)-2-(methoxymethyl)pyrrolidin-1-amine
SMILES COCC1CCCN1N

Specifications

Color Yellow
Boiling Point 64°C
Quantity 1 g
Formula Weight 130.19
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material (S)-(-)-1-Amino-2-(methoxymethyl)pyrrolidine