missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-(4-Methylphenyl)ethylamine, ChiPros 98%, ee 99+%

Catalog No. AAL1632914
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1632914 25 g
AAL1632903 1 g
AAL1632906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1632914 Supplier Thermo Scientific Chemicals Supplier No. L1632914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 27298-98-2 | C9H13N | 135.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27298-98-2
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00145246
InChI Key UZDDXUMOXKDXNE-UHFFFAOYNA-N
Synonym s-1-p-tolyl ethanamine, s---1-p-tolyl ethylamine, s---4-1-aminoethyl toluene, s-1-p-tolyl ethylamine, 1s-1-4-methylphenyl ethan-1-amine, 1s-1-p-tolyl ethanamine, 1s-1-4-methylphenyl ethanamine, s-1-p-tolylethanamine, 1-4-methylphenyl ethanamine #, s-1-4-methylphenyl ethylamine
PubChem CID 7015759
SMILES CC(N)C1=CC=C(C)C=C1

Specifications

Melting Point <-20°C
Density 0.94
Boiling Point 204°C
Flash Point 82°C (179°F)
Refractive Index 1.5215
Quantity 25 g
UN Number UN2735
Beilstein 2422644
Sensitivity Air Sensitive
IUPAC Name (1S)-1-(4-methylphenyl)ethanamine
Formula Weight 135.21
Percent Purity 98%
Assay ee ≥99%
Chemical Name or Material (S)-(-)-1-(4-Methylphenyl)ethylamine
Show More Show Less
Hazard Category H227-H302-H314-H317-H335
Hazard Statement GHS H Statement
H314-H318-H302-H312-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 623-635-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only