missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-(4-Chlorophenyl)ethylamine, ChiPros™ 94%

CAS: 4187-56-8 | C8H10ClN | 155.625 g/mol

$73.42 - $686.70

Chemical Identifiers

CAS 4187-56-8
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.625
MDL Number MFCD00671640
InChI Key PINPOEWMCLFRRB-LURJTMIESA-N
Synonym s-1-4-chlorophenyl ethylamine, s-1-4-chloro-phenyl ethylamine, 1s-1-4-chlorophenyl ethanamine, 1s-1-4-chlorophenyl ethanamine, s---1-4-chlorophenyl ethylamine, s---1-4-chlorophenyl ethylamine, pubchem15221, s-4-chloro-, a-methylbenzylamine, 1s-1-4-chlorophenyl ethylamine, 1s-1-4-chlorophenyl ethylamine
PubChem CID 1550852
IUPAC Name (1S)-1-(4-chlorophenyl)ethanamine
SMILES CC(C1=CC=C(C=C1)Cl)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6671503
View Documents
Thermo Scientific Chemicals
H6671503
1 g
Each for $73.42
Only null left
 
AAH6671506
View Documents
Thermo Scientific Chemicals
H6671506
5 g
Each for $190.66
Only null left
 
AAH6671514
View Documents
Thermo Scientific Chemicals
H6671514
25 g
Each for $686.70
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4187-56-8
155.625
PINPOEWMCLFRRB-LURJTMIESA-N
1550852
CC(C1=CC=C(C=C1)Cl)N
C8H10ClN
MFCD00671640
s-1-4-chlorophenyl ethylamine, s-1-4-chloro-phenyl ethylamine, 1s-1-4-chlorophenyl ethanamine, 1s-1-4-chlorophenyl ethanamine, s---1-4-chlorophenyl ethylamine, s---1-4-chlorophenyl ethylamine, pubchem15221, s-4-chloro-, a-methylbenzylamine, 1s-1-4-chlorophenyl ethylamine, 1s-1-4-chlorophenyl ethylamine
(1S)-1-(4-chlorophenyl)ethanamine

Specifications

4187-56-8
1.110g/mL
1 g
Pungent
1.5418
MFCD00671640
2412316
PINPOEWMCLFRRB-LURJTMIESA-N
(1S)-1-(4-chlorophenyl)ethanamine
1550852
94%
Yellow
232°C
115°C (239°F)
Liquid
C8H10ClN
UN2922
s-1-4-chlorophenyl ethylamine, s-1-4-chloro-phenyl ethylamine, 1s-1-4-chlorophenyl ethanamine, 1s-1-4-chlorophenyl ethanamine, s-4-chloro-alpha-methylbenzylamine, s---1-4-chlorophenyl ethylamine, pubchem15221, pubchem15221, a-methylbenzylamine, 1s-1-4-chlorophenyl ethylamine, 1s-1-4-chlorophenyl ethylamine
CC(C1=CC=C(C=C1)Cl)N
155.625
155.63
(S)-(-)-1-(4-Chlorophenyl)ethylamine, ChiPros™
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H330-H314-H318
Toxic if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P272-P280-P284-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P330-P333+P313-P362-P501c

H302-H315-H317-H318-H330

DOTInformation : Hazard Class: 6.1; Packaging Group: II

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only