missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-(4-Chlorophenyl)ethylamine, ChiPros™ 94%

Catalog No. AAH6671514
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6671514 25 g
AAH6671503 1 g
AAH6671506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6671514 Supplier Thermo Scientific Chemicals Supplier No. H6671514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4187-56-8 | C8H10ClN | 155.625 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4187-56-8
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.625
MDL Number MFCD00671640
InChI Key PINPOEWMCLFRRB-LURJTMIESA-N
Synonym s-1-4-chlorophenyl ethylamine, s-1-4-chlorophenyl ethanamine, 1s-1-4-chlorophenyl ethanamine, s-4-chloro-alpha-methylbenzylamine, s---1-4-chlorophenyl ethylamine, 1s-1-4-chlorophenyl ethan-1-amine, pubchem15221, s-4-chloro-, a-methylbenzylamine, 1s-1-4-chlorophenyl ethylamine, s-1-4-chloro-phenyl ethylamine
PubChem CID 1550852
IUPAC Name (1S)-1-(4-chlorophenyl)ethanamine
SMILES CC(C1=CC=C(C=C1)Cl)N

Specifications

Color Yellow
Density 1.110g/mL
Quantity 25 g
Flash Point 115°C (239°F)
Odor Pungent
Physical Form Liquid
Refractive Index 1.5418
UN Number UN2922
Beilstein 2412316
Boiling Point 232°C
Formula Weight 155.63
Percent Purity 94%
Chemical Name or Material (S)-(-)-1-(4-Chlorophenyl)ethylamine, ChiPros™
Show More Show Less
Hazard Category H302-H315-H317-H318-H330
Hazard Statement GHS H Statement
H301-H330-H314-H318
Toxic if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P272-P280-P284-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P330-P333+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only