missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-1,3-Butanediol 98.0+%, TCI America™
$109.74 - $379.26
Chemical Identifiers
| CAS | 24621-61-2 |
|---|---|
| Molecular Formula | C4H10O2 |
| Molecular Weight (g/mol) | 90.122 |
| MDL Number | MFCD00064278 |
| InChI Key | PUPZLCDOIYMWBV-BYPYZUCNSA-N |
| Synonym | s-+-1,3-butanediol, s-butane-1,3-diol, 3s-butane-1,3-diol, s-1,3-butanediol, s-+-butane-1,3-diol, 1,3-butanediol, 3s, bu2, 1,3-butanediol, s, pubchem6722 |
| PubChem CID | 446973 |
| ChEBI | CHEBI:52688 |
| IUPAC Name | (3S)-butane-1,3-diol |
| SMILES | CC(CCO)O |
Chemical Identifiers
| 24621-61-2 | |
| 90.122 | |
| PUPZLCDOIYMWBV-BYPYZUCNSA-N | |
| 446973 | |
| (3S)-butane-1,3-diol |
| C4H10O2 | |
| MFCD00064278 | |
| s-+-1,3-butanediol, s-butane-1,3-diol, 3s-butane-1,3-diol, s-1,3-butanediol, s-+-butane-1,3-diol, 1,3-butanediol, 3s, bu2, 1,3-butanediol, s, pubchem6722 | |
| CHEBI:52688 | |
| CC(CCO)O |
Specifications
| 24621-61-2 | |
| 109°C | |
| MFCD00064278 | |
| s-+-1,3-butanediol, s-butane-1,3-diol, 3s-butane-1,3-diol, s-1,3-butanediol, s-+-butane-1,3-diol, 1,3-butanediol, 3s, bu2, 1,3-butanediol, s, pubchem6722 | |
| CC(CCO)O | |
| 90.122 | |
| CHEBI:52688 | |
| ≥98.0% (GC) | |
| (S)-(+)-1,3-Butanediol |
| Yellow | |
| C4H10O2 | |
| 1 g | |
| PUPZLCDOIYMWBV-BYPYZUCNSA-N | |
| (3S)-butane-1,3-diol | |
| 446973 | |
| 90.12 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0235
TSCA : No