missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1,3-Butanediol 98.0+%, TCI America™

Catalog No. B11605G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11605G 5 g
B11601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B11605G Supplier TCI America Supplier No. B11605G

Chemical Identifiers

CAS 24621-61-2
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.122
MDL Number MFCD00064278
InChI Key PUPZLCDOIYMWBV-BYPYZUCNSA-N
Synonym s-+-1,3-butanediol, s-butane-1,3-diol, 3s-butane-1,3-diol, s-1,3-butanediol, s-+-butane-1,3-diol, 1,3-butanediol, 3s, bu2, 1,3-butanediol, s, pubchem6722
PubChem CID 446973
ChEBI CHEBI:52688
IUPAC Name (3S)-butane-1,3-diol
SMILES CC(CCO)O

Specifications

Color Yellow
Boiling Point 109°C
Quantity 5 g
Formula Weight 90.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-(+)-1,3-Butanediol
EINECSNumber (2)-0235
TSCA No