missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-1,3-Butanediol, 98%
CAS: 24621-61-2 | C4H10O2 | 90.12 g/mol
$185.91 - $185.91
Chemical Identifiers
| CAS | 24621-61-2 |
|---|---|
| Molecular Formula | C4H10O2 |
| Molecular Weight (g/mol) | 90.12 |
| MDL Number | MFCD00064278 |
| InChI Key | PUPZLCDOIYMWBV-BYPYZUCNSA-N |
| Synonym | s-+-1,3-butanediol, s-butane-1,3-diol, 3s-butane-1,3-diol, s-1,3-butanediol, s-+-butane-1,3-diol, 1,3-butanediol, 3s, bu2, 1,3-butanediol, s, pubchem6722 |
| PubChem CID | 446973 |
| ChEBI | CHEBI:52688 |
| IUPAC Name | (3S)-butane-1,3-diol |
| SMILES | CC(CCO)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC217970010
|
Thermo Scientific Chemicals
217970010 |
1 g | Glass bottle |
Each for $185.91
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 24621-61-2 | |
| 90.12 | |
| PUPZLCDOIYMWBV-BYPYZUCNSA-N | |
| 446973 | |
| (3S)-butane-1,3-diol |
| C4H10O2 | |
| MFCD00064278 | |
| s-+-1,3-butanediol, s-butane-1,3-diol, 3s-butane-1,3-diol, s-1,3-butanediol, s-+-butane-1,3-diol, 1,3-butanediol, 3s, bu2, 1,3-butanediol, s, pubchem6722 | |
| CHEBI:52688 | |
| CC(CCO)O |
Specifications
| 24621-61-2 | |
| 100.0 | |
| 1.0000g/mL | |
| 121°C | |
| 97.5% min. (GC) | |
| C4H10O2 | |
| CH3CH(OH)CH2CH2OH | |
| 1 g | |
| + 30.00 | |
| Solubility in water: soluble. Other solubilities: soluble in acetone, methyl ethyl ketone, ethanol, slightly soluble in ether, practically insoluble in aliphatic hydrocarbons,, benzene, toluene and ccl4 | |
| CC(CCO)O | |
| 90.12 | |
| CHEBI:52688 | |
| 98% | |
| (S)-(+)-1, 3-Butanediol |
| 97.5 | |
| Colorless to Yellow | |
| 109.0°C (14.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4385 to 1.4405 | |
| MFCD00064278 | |
| 1 | |
| s-+-1,3-butanediol, s-butane-1,3-diol, 3s-butane-1,3-diol, s-1,3-butanediol, s-+-butane-1,3-diol, 1,3-butanediol, 3s, bu2, 1,3-butanediol, s, pubchem6722 | |
| PUPZLCDOIYMWBV-BYPYZUCNSA-N | |
| (3S)-butane-1,3-diol | |
| 446973 | |
| 90.12 | |
| Viscous Liquid |
Safety and Handling
EINECSNumber : 246-363-
RUO – Research Use Only