missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-1,2-Propanediol 98.0+%, TCI America™
Supplier: TCI America P112925G
Specifications
| (S)-(+)-1,2-Propanediol | |
| Yellow | |
| C3H8O2 | |
| 25 g | |
| DNIAPMSPPWPWGF-UHFFFAOYNA-N | |
| propane-1,2-diol | |
| 439846 | |
| 76.10 | |
| Liquid |
| 4254-15-3 | |
| 92°C | |
| MFCD00004539 | |
| s-+-1,2-propanediol, s-propane-1,2-diol, s-1,2-propanediol, 2s-propane-1,2-diol, s-propylene glycol, s-+-propylene glycol, s-+-1,2-dihydroxypropane, propylene glycol #, pgo | |
| CC(O)CO | |
| 76.10 | |
| CHEBI:29002 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 4254-15-3 | |
| 76.10 | |
| DNIAPMSPPWPWGF-UHFFFAOYNA-N | |
| 439846 | |
| propane-1,2-diol |
| C3H8O2 | |
| MFCD00004539 | |
| s-+-1,2-propanediol, s-propane-1,2-diol, s-1,2-propanediol, 2s-propane-1,2-diol, s-propylene glycol, s-+-propylene glycol, s-+-1,2-dihydroxypropane, propylene glycol #, pgo | |
| CHEBI:29002 | |
| CC(O)CO |
Safety and Handling
EINECSNumber : (2)-0234
TSCA : No