missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1,2-Propanediol 98.0+%, TCI America™

Catalog No. P11295G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11295G 5 g
P112925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11295G Supplier TCI America Supplier No. P11295G

Chemical Identifiers

CAS 4254-15-3
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.10
MDL Number MFCD00004539
InChI Key DNIAPMSPPWPWGF-UHFFFAOYNA-N
Synonym s-+-1,2-propanediol, s-propane-1,2-diol, s-1,2-propanediol, 2s-propane-1,2-diol, s-propylene glycol, s-+-propylene glycol, s-+-1,2-dihydroxypropane, propylene glycol #, pgo
PubChem CID 439846
ChEBI CHEBI:29002
IUPAC Name propane-1,2-diol
SMILES CC(O)CO

Specifications

Color Yellow
Boiling Point 92°C
Quantity 5 g
Formula Weight 76.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-(+)-1,2-Propanediol
EINECSNumber (2)-0234
TSCA No