missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-(2-Naphthyl)ethylamine 98.0+%, TCI America™

Catalog No. p-7133953
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N07261G 1 g
N07265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N07261G Supplier TCI America Supplier No. N07261G

Chemical Identifiers

CAS 3082-62-0
Molecular Formula C12H13N
Molecular Weight (g/mol) 171.243
MDL Number MFCD00085366
InChI Key KHSYYLCXQKCYQX-VIFPVBQESA-N
Synonym s-1-naphthalen-2-yl ethanamine, s---1-2-naphthyl ethylamine, s-1-2-naphthyl ethylamine, 1s-1-naphthalen-2-yl ethan-1-amine, 2-naphthalenemethanamine, alpha-methyl-, alphas, 1s-1-2-naphthyl ethylamine, 1s-1-naphthalen-2-yl ethanamine, s-1-2-naphthyl ethanamine, s-1-naphth-2-yl ethylamine, s-1-naphth-2-yl-ethylamine
PubChem CID 1201503
IUPAC Name (1S)-1-naphthalen-2-ylethanamine
SMILES CC(C1=CC2=CC=CC=C2C=C1)N

Specifications

Melting Point 53°C
Color White-Yellow
Boiling Point 143°C
Quantity 1 g
Formula Weight 171.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-1-(2-Naphthyl)ethylamine