missing translation for 'onlineSavingsMsg'
Learn More

(S)-1,2-Diaminopropane Dihydrochloride 98.0+%, TCI America™

Catalog No. D28281G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D28281G Supplier TCI America Supplier No. D28281G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19777-66-3
Molecular Formula C3H12Cl2N2
Molecular Weight (g/mol) 147.043
MDL Number MFCD00050706
InChI Key AEIAMRMQKCPGJR-QTNFYWBSSA-N
Synonym s---1,2-diaminopropane dihydrochloride, s-propane-1,2-diamine dihydrochloride, s-1,2-diaminopropane dihydrochloride, s---propylenediamine dihydrochloride, s-1,2-propanediamine dihydrochloride, 2s-propane-1,2-diamine dihydrochloride, s-1,2-diaminopropane 2hcl, s-propylenediamine dihydrochloride, 1,2-propanediamine, dihydrochloride, s, pubchem15232
PubChem CID 12217445
IUPAC Name (2S)-propane-1,2-diamine;dihydrochloride
SMILES CC(CN)N.Cl.Cl

Specifications

Color White
Quantity 1 g
Formula Weight 147.04
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-1,2-Diaminopropane Dihydrochloride
EINECSNumber (2)-0149
TSCA No