missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-(2-Chlorophenyl)ethanol, 98%

Catalog No. aah6093503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6093503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6093503 Supplier Thermo Scientific Chemicals Supplier No. H6093503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 131864-71-6 | C8H9ClO | 156.609 g/mol

It is an intermediate in organic syntheses and used as a pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an intermediate in organic syntheses and used as a pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents. Store under room tempera

Chemical Identifiers

CAS 131864-71-6
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.609
MDL Number MFCD06659506
InChI Key DDUBOVLGCYUYFX-LURJTMIESA-N
Synonym s-1-2-chlorophenyl ethanol, 1s-1-2-chlorophenyl ethan-1-ol, 1s-1-2-chlorophenyl ethanol, s-1-2-chlorophenyl ethan-1-ol, pubchem5670, s-1-2-chloro-phenyl-ethanol, 5-1-2-chloro-phenyl-ethanol, s-2-chloro-alpha-methylbenzyl alcohol, benzenemethanol,2-chloro-a-methyl-, as
PubChem CID 6999091
IUPAC Name (1S)-1-(2-chlorophenyl)ethanol
SMILES CC(C1=CC=CC=C1Cl)O

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 156.61
Percent Purity 98%
Chemical Name or Material (S)-1-(2-Chlorophenyl)ethanol
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only