missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-(1-Naphthyl)ethylamine 99.0+%, TCI America™

Catalog No. p-7133831
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N04811G 1 g
N04815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N04811G Supplier TCI America Supplier No. N04811G

Chemical Identifiers

CAS 10420-89-0
Molecular Formula C12H14N
Molecular Weight (g/mol) 172.25
MDL Number MFCD00064179
InChI Key RTCUCQWIICFPOD-VIFPVBQESA-O
Synonym s---1-1-naphthyl ethylamine, s-1-naphthalen-1-yl ethanamine, 1s-1-naphthalen-1-yl ethan-1-amine, s---alpha-1-naphthyl ethylamine, s-1-1-naphthyl ethylamine, 1s-1-1-naphthyl ethanamine, s---1-naphthyl ethylamine, s-alpha-methyl-1-naphthalenemethanamine, 1s-1-naphthalen-1-yl ethanamine
PubChem CID 66325
IUPAC Name (1S)-1-(naphthalen-1-yl)ethan-1-aminium
SMILES C[C@H]([NH3+])C1=C2C=CC=CC2=CC=C1

Specifications

Color Yellow
Boiling Point 125°C
Quantity 1 g
Formula Weight 171.24
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-(-)-1-(1-Naphthyl)ethylamine
EINECSNumber (4)-1343
RTECSNumber QJ6963000
TSCA Yes
Recommended Storage Refrigerator