missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1,1'-Binaphthyl-2,2'-diyl Bis(trifluoromethanesulfonate) 93.0+%, TCI America™

Supplier:  TCI America B22311G

 View more versions of this product

Catalog No. B22311G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

(S)-(+)-1,1′-Binaphthyl-2,2′-diyl Bis(trifluoromethanesulfonate)
83°C
C22H12F6O6S2
1 g
r-1,1'-binaphthalene-2,2'-diyl bis trifluoromethanesulfonate, s-1,1'-binaphthalene-2,2'-diyl bis trifluoromethanesulfonate, r---1,1'-bi-2-naphthol bis trifluoromethanesulfonate, r---1,1'-binaphthol-2,2'-bis trifluoromethanesulfonate, s-+-1,1'-bi-2-naphthol bis trifluoromethanesulfonate, r-1,1'-binaphthyl-2,2'-diyl bis trifluoromethane sulphonate, s-+-1,1'-binaphthol-2,2'-bis trifluoromethanesulfonate, 1,1'-bi-2-naphthol bis trifluoromethanesulfonate, s-1,1'-bi 2-naphthol bis trifluoromethanesulfonate, r---binol-tf2
C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)OS(=O)(=O)C(F)(F)F)OS(=O)(=O)C(F)(F)F
550.442
550.44
Crystalline Powder
128544-05-8
White-Yellow
MFCD00274615
3261
OYJLCOSEYYZULE-UHFFFAOYSA-N
[1-[2-(trifluoromethylsulfonyloxy)naphthalen-1-yl]naphthalen-2-yl] trifluoromethanesulfonate
389883
≥93.0% (HPLC)

Chemical Identifiers

128544-05-8
550.442
OYJLCOSEYYZULE-UHFFFAOYSA-N
389883
C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)OS(=O)(=O)C(F)(F)F)OS(=O)(=O)C(F)(F)F
C22H12F6O6S2
MFCD00274615
r-1,1'-binaphthalene-2,2'-diyl bis trifluoromethanesulfonate, s-1,1'-binaphthalene-2,2'-diyl bis trifluoromethanesulfonate, r---1,1'-bi-2-naphthol bis trifluoromethanesulfonate, r---1,1'-binaphthol-2,2'-bis trifluoromethanesulfonate, s-+-1,1'-bi-2-naphthol bis trifluoromethanesulfonate, r-1,1'-binaphthyl-2,2'-diyl bis trifluoromethane sulphonate, s-+-1,1'-binaphthol-2,2'-bis trifluoromethanesulfonate, 1,1'-bi-2-naphthol bis trifluoromethanesulfonate, s-1,1'-bi 2-naphthol bis trifluoromethanesulfonate, r---binol-tf2
[1-[2-(trifluoromethylsulfonyloxy)naphthalen-1-yl]naphthalen-2-yl] trifluoromethanesulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions