missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ruthenium Red, ≥85%, pure

Catalog No. AC279710010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC279710010 1 g
AC279710050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC279710010 Supplier Thermo Scientific Chemicals Supplier No. 279710010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 11103-72-3 | H24Cl6N14O2Ru3 | 786.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 11103-72-3
Molecular Formula H24Cl6N14O2Ru3
Molecular Weight (g/mol) 786.35
InChI Key JQJSTVUROJELSR-UHFFFAOYSA-H
Synonym 6cl.o2ru3.14nh3, ruthenium red, technical grade, degrees +/->> aeno>> ie, azane;ruthenium 2+ ;hexachloride;dihydrate, ruthenium red, for microscopy calc. on dry substance, at, ruthenium red, for microscopy calc. on dry basis, at, triruthenoxane-1,1,3,3,5,5-hexakis ylium tetradecaamine hexachloride
PubChem CID 16218584
IUPAC Name azane;ruthenium(2+);hexachloride;dihydrate
SMILES N.N.N.N.N.N.N.N.N.N.N.N.N.N.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru+2].[Ru+2].[Ru+2]

Specifications

Melting Point > 500.0°C
Quantity 1 g
Assay Percent Range ≥85%
Linear Formula [(NH3)5RuORu(NH3)4ORu(NH3)5]Cl6
Merck Index 15,8434
Solubility Information Solubility in water: soluble.
Formula Weight 786.35
Percent Purity ≥85%
Grade Pure
Packaging Glass Bottle
Chemical Name or Material Ruthenium Red
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protec

RUO – Research Use Only