Learn More
Ruthenium(IV) oxide hydrate, Premion™, 99.99% (metals basis), Ru 54-60%
CAS: 32740-79-7 | O2Ru | 133.07 g/mol
$165.58 - $165.58
Chemical Identifiers
| CAS | 32740-79-7 |
|---|---|
| Molecular Formula | O2Ru |
| Molecular Weight (g/mol) | 133.07 |
| MDL Number | MFCD00149846 |
| InChI Key | WOCIAKWEIIZHES-UHFFFAOYSA-N |
| IUPAC Name | dioxoruthenium |
| SMILES | O=[Ru]=O |
Description
Chemical catalyst, important material for producing resistance and capacitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 32740-79-7 | |
| 133.07 | |
| WOCIAKWEIIZHES-UHFFFAOYSA-N | |
| O=[Ru]=O |
| O2Ru | |
| MFCD00149846 | |
| dioxoruthenium |
Specifications
| 1200°C (sublimation) | |
| Black | |
| 1 g | |
| MFCD00149846 | |
| It is soluble in fused alkalis. It is insoluble in water. | |
| O=[Ru]=O | |
| 133.07 | |
| 99.99% | |
| Ru 54 to 60% | |
| (metals basis) |
| 32740-79-7 | |
| Powder | |
| O2Ru | |
| UN1479 | |
| WOCIAKWEIIZHES-UHFFFAOYSA-N | |
| dioxoruthenium | |
| 133.07 (Anhydrous) | |
| Premion | |
| Ruthenium (IV) oxide hydrate |
Safety and Handling
GHS H Statement
H271
May cause fire or explosion
strong oxidizer.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P501c
H315-H319
DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
EINECSNumber : 234-840-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only