missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(III) chloride oxide, ammoniated

Catalog No. AA1180603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1180603 1 g
AA1180606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1180603 Supplier Thermo Scientific Chemicals Supplier No. 01180603

Microscopic stain | CAS: 11103-72-3 | Cl6H46N14O2Ru3 | 790.374 g/mol

Microscopic stainRuthenium(III) chloride oxide, ammoniated is used as a reagent on plant material for staining pectin, gum, animal tissues and bacteria. It is also used in histology to stain aldehyde fixed mucopolysaccharides. Further, it serves as a pharmacological tool to study specific cellular mechanisms. It serves as an inhibitor of mitochondrial calcium uniporter. In addition, it is involved in reducing airway inflammation arises by pepper spray.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 11103-72-3
Molecular Formula Cl6H46N14O2Ru3
Molecular Weight (g/mol) 790.374
MDL Number MFCD00011479
InChI Key JQJSTVUROJELSR-UHFFFAOYSA-H
Synonym 6cl.o2ru3.14nh3, ruthenium red, technical grade, degrees +/->> aeno>> ie, azane;ruthenium 2+ ;hexachloride;dihydrate, ruthenium red, for microscopy calc. on dry substance, at, ruthenium red, for microscopy calc. on dry basis, at, triruthenoxane-1,1,3,3,5,5-hexakis ylium tetradecaamine hexachloride
PubChem CID 16218584
IUPAC Name azane;ruthenium(2+);hexachloride;dihydrate
SMILES N.N.N.N.N.N.N.N.N.N.N.N.N.N.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru+2].[Ru+2].[Ru+2]

Specifications

Melting Point >500°C (decomposition)
Color Red
Physical Form Crystalline
Quantity 1 g
Linear Formula [(NH3)5RuORu(NH3)4ORu(NH3)5]Cl6
Merck Index 14,8300
Solubility Information Soluble in water and ammonium hydroxide.
Formula Weight 786.35
Concentration or Composition (by Analyte or Components) ≥34.8% Ruthenium
Odor Odorless
Density 3.110 g/mL
Chemical Name or Material Ruthenium(III) chloride oxide, Ammoniated
Show More Show Less
Hazard Category H315-H319
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
RTECSNumber VM2656500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only