missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(III) bromide hydrate, Ru 25% min

Catalog No. p-7049060
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1262803 1 g
AA1262806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1262803 Supplier Thermo Scientific Chemicals Supplier No. 01262803
Only null left

Oxidation | CAS: 314075-07-5 | Br3H2ORu | 358.80 g/mol

Ruthenium(III) bromide hydrate is used as a catalyst in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 314075-07-5
Molecular Formula Br3H2ORu
Molecular Weight (g/mol) 358.80
MDL Number MFCD00016314
InChI Key PMMMCGISKBNZES-UHFFFAOYSA-K
IUPAC Name tribromoruthenium hydrate
SMILES O.Br[Ru](Br)Br

Specifications

Physical Form Crystalline
Quantity 1 g
UN Number UN3260
Solubility Information Soluble in alcohol
Formula Weight 340.78 (Anhydrous)
Concentration or Composition (by Analyte or Components) Ru 25% min.
Odor Characteristic
Sensitivity Hygroscopic
Chemical Name or Material Ruthenium(III) bromide hydrate
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 237-829-4
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only