missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bolle Safety™ Rush+ Safety Glasses
Rush+ safety spectacles eco pack.
$370.71 - $370.71
Specifications
| Lens Coating | Anti-fog, Scratch-resistant |
|---|---|
| Product Type | Safety Glasses |
| Material | Polycarbonate |
| Disposable | Reusable |
| Lens Material | Polycarbonate |
| Catalog Number | Mfr. No. | Color | Lens Tint | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Lens Tint | Price | Quantity | ||||
|
17-623-366
|
Bolle Safety™
PSSRUSP073 |
Black with Red Frame | Clear |
Pack of 20 Each for $370.71
|
|
||||
Description
Rush+ safety spectacles eco pack, box of 20.
Specifications
| Anti-fog, Scratch-resistant | |
| Polycarbonate | |
| Polycarbonate | |
| None | |
| Not Autoclavable |
| Safety Glasses | |
| Reusable | |
| Polycarbonate | |
| Standard |