missing translation for 'onlineSavingsMsg'
Learn More

Rufinamide 98.0+%, TCI America™

Catalog No. R014325MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R014325MG 25 mg
R0143100MG 100 mg
R01431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R014325MG Supplier TCI America Supplier No. R014325MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 106308-44-5
Molecular Formula C10H8F2N4O
Molecular Weight (g/mol) 238.198
MDL Number MFCD00865314
InChI Key POGQSBRIGCQNEG-UHFFFAOYSA-N
Synonym rufinamide, inovelon, banzel, 1-2,6-difluorobenzyl-1h-1,2,3-triazole-4-carboxamide, xilep, 1-2,6-difluorophenyl methyl-1h-1,2,3-triazole-4-carboxamide, unii-wfw942pr79, 1-2,6-difluorophenyl methyl triazole-4-carboxamide, 1h-1,2,3-triazole-4-carboxamide, 1-2,6-difluorophenyl methyl, dsstox_cid_26506
PubChem CID 129228
IUPAC Name 1-[(2,6-difluorophenyl)methyl]triazole-4-carboxamide
SMILES C1=CC(=C(C(=C1)F)CN2C=C(N=N2)C(=O)N)F

Specifications

Melting Point 241°C
Color White
Quantity 25 mg
Formula Weight 238.20
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Rufinamide