missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RuCl2[(S)-xylbinap][(S)-daipen], TCI America™
Specifications
Specifications
| Quantity | 200 mg |
| Formula Weight | 1221.30 |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | RuCl2[(S)-xylbinap][(S)-daipen] |
Safety and Handling
| TSCA | No |