missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RuCl2[(R)-xylbinap][(R)-daipen], TCI America™
$139.81 - $475.33
Chemical Identifiers
| CAS | 220114-32-9 |
|---|---|
| Molecular Formula | C71H74Cl2N2O2P2Ru |
| MDL Number | MFCD09753026 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
R0133200MG
|
TCI America
R0133200MG |
200 mg |
Each for $139.81
|
|
|||||
|
R01331G
|
TCI America
R01331G |
1 g |
Each for $475.33
|
|
|||||
Chemical Identifiers
| 220114-32-9 | |
| C71H74Cl2N2O2P2Ru | |
| MFCD09753026 |
Specifications
| 220114-32-9 | |
| MFCD09753026 | |
| 1221.30 | |
| RuCl2[(R)-xylbinap][(R)-daipen] |
| C71H74Cl2N2O2P2Ru | |
| 200 mg | |
| Crystalline Powder |
Safety and Handling
TSCA : No