missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RuCl[(R,R)-Fsdpen](p-cymene) 90.0+%, TCI America™
$141.67 - $495.84
Chemical Identifiers
| CAS | 1026995-71-0 |
|---|---|
| Molecular Formula | C30H28ClF5N2O2RuS |
| Molecular Weight (g/mol) | 712.138 |
| MDL Number | MFCD12545963 |
| InChI Key | UWFMZLATRGEOIW-ZJPTYAPPSA-M |
| Synonym | [(R,R)-N-(2-Amino-1,2-diphenylethyl)pentafluorobenzenesulfonamide]chloro(p-cymene)ruthenium(II), Chloro(p-cymene)[(R,R)-N-(pentafluorobenzenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II) |
| PubChem CID | 71463829 |
| IUPAC Name | [(1R,2R)-2-amino-1,2-diphenylethyl]-(2,3,4,5,6-pentafluorophenyl)sulfonylazanide;1-methyl-4-propan-2-ylbenzene;ruthenium(2+);chloride |
| SMILES | CC1=CC=C(C=C1)C(C)C.C1=CC=C(C=C1)C(C(C2=CC=CC=C2)[N-]S(=O)(=O)C3=C(C(=C(C(=C3F)F)F)F)F)N.[Cl-].[Ru+2] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
R0123200MG
|
TCI America
R0123200MG |
200 mg |
Each for $141.67
|
|
|||||
|
R01231G
|
TCI America
R01231G |
1 g |
Each for $495.84
|
|
|||||
Chemical Identifiers
| 1026995-71-0 | |
| 712.138 | |
| UWFMZLATRGEOIW-ZJPTYAPPSA-M | |
| 71463829 | |
| CC1=CC=C(C=C1)C(C)C.C1=CC=C(C=C1)C(C(C2=CC=CC=C2)[N-]S(=O)(=O)C3=C(C(=C(C(=C3F)F)F)F)F)N.[Cl-].[Ru+2] |
| C30H28ClF5N2O2RuS | |
| MFCD12545963 | |
| [(R,R)-N-(2-Amino-1,2-diphenylethyl)pentafluorobenzenesulfonamide]chloro(p-cymene)ruthenium(II), Chloro(p-cymene)[(R,R)-N-(pentafluorobenzenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II) | |
| [(1R,2R)-2-amino-1,2-diphenylethyl]-(2,3,4,5,6-pentafluorophenyl)sulfonylazanide;1-methyl-4-propan-2-ylbenzene;ruthenium(2+);chloride |
Specifications
| 1026995-71-0 | |
| C30H28ClF5N2O2RuS | |
| 200 mg | |
| UWFMZLATRGEOIW-ZJPTYAPPSA-M | |
| [(1R,2R)-2-amino-1,2-diphenylethyl]-(2,3,4,5,6-pentafluorophenyl)sulfonylazanide;1-methyl-4-propan-2-ylbenzene;ruthenium(2+);chloride | |
| 71463829 | |
| ≥90% (NMR) | |
| RuCl[(R,R)-Fsdpen](p-cymene) |
| 244°C | |
| MFCD12545963 | |
| [(R,R)-N-(2-Amino-1,2-diphenylethyl)pentafluorobenzenesulfonamide]chloro(p-cymene)ruthenium(II), Chloro(p-cymene)[(R,R)-N-(pentafluorobenzenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II) | |
| CC1=CC=C(C=C1)C(C)C.C1=CC=C(C=C1)C(C(C2=CC=CC=C2)[N-]S(=O)(=O)C3=C(C(=C(C(=C3F)F)F)F)F)N.[Cl-].[Ru+2] | |
| 712.138 | |
| 712.14 | |
| Crystalline Powder |
Safety and Handling
TSCA : No